Mouse IgG2b Isotype Control [clone: MPC-11]
Catalog # 70-4732-U100
Supplier: Tonbo Biosciences
Specifications
- Antibody type:Control
- Clone:MPC-11
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Storage temperature:Store undiluted at 2-8 °C, protect from light, do not freeze.
- Size:100 μg
- Pk:100 µG
Specifications
About this item
The MPC-11 immunoglobulin is useful as an isotype-matched control. The MPC-11 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG2b antibodies.
Type: Control
Antigen: MPC-11
Clonality:
Clone: MPC-11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: