About this item
AQUAL are designed for the preparation of high quality pure (demi) water, which is used for analytical and technical purposes. All types of devices consist of filters, which are used to remove coarse and fine mechanical impurities and dechlorination, and a reverse osmosis module, which removes a substantial part of dissolved general salts and other impurities by at least 96% of the incoming water.
In the basic version, an ion exchange resin of 0.75 l is used as the finishing stage for devices of the type series starting with 3 (for example 35). In this variant, it is necessary to take into account more frequent filling changes. An integral part of AQUAL devices can be, for example, a digital conductivity meter, UV lamp, bacterial filter, pump for increasing the inlet pressure, electromagnetic triggering of the device in combination with a storage tank or also central distribution of treated water.
AQUALU 25-29-1 without demicolone, only the percentage desalination is given because the input conductivity is not known in advance and therefore the resulting conductivity cannot be determined.
AQAULU 29-29-2 with demicolone shows the quality of the resulting water in uS / cm because it is relatively one (according to the input conductivity) what is the value of conductivity behind reverse osmosis, because the remaining part is purified by the just included demineralization column. For this reason, we guarantee a value below 1 uS / cm. The demineralization column is able to adjust the water from any input conductivity (normal) to the previously mentioned values. the output depends on the inlet water pressure and temperature.
The AQUAL range of devices continues with 40, 80, 100, 150, 300 and 600 l / h devices.