Anti-NUP43 Rabbit Polyclonal Antibody
Catalog # ABGEAP17331C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:nucleoporin 43kDa
- Antigen symbol:NUP43
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:internal
- Gene ID:348995
- Antigen synonyms:bA350J20.1|p42
- Amino acid number:142 to 171
- Storage buffer:pH 7,4
- Molecular weight:42.151 kDa
- Sequence:RADHKEAVRTIDNADC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.43
- Shipping temperature:4 °C
- Immunogen:This NUP43 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 142-171 amino acids from the Central region of human NUP43.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: NUP43
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human