Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-PPARA Mouse Monoclonal Antibody [clone: 1331CT894.186.143]
Katalogové číslo ABGEAM8425B
Dodavatel:  Abgent
undefined
Anti-PPARA Mouse Monoclonal Antibody [clone: 1331CT894.186.143]
Katalogové číslo ABGEAM8425B
Dodavatel:  Abgent

Specifikace

  • Antibody type:
    Primary
  • Antigen name:
    peroxisome proliferator-activated receptor alpha
  • Antigen symbol:
    PPARA
  • Clonality:
    Monoclonal
  • Clone:
    1331CT894.186.143
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Gene ID:
    5465
  • Antigen synonyms:
    PPAR|PPARalpha|NR1C1|hPPAR
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    52.225 kDa
  • Sequence:
    MVDTESPLCPLSPLEAGDLESPLSEEFLQEMGNIQEISQSIGEDSSGSFGFTEYQYLGSCPGSDGSVITDTLSPASSPSSVTYPVVPGSVDESPSGALNIECRICGDKASGYHYGVHACEGCKGFFRRTIRLKLVYDKCDRSCKIQKKNRNKCQYCRFHKCLSVGMSHNAIRFGRMPRSEKAKLKAEILTCEHDIEDSETADLKSLAKRIYEAYLKNFNMNKVKARVILSGKASNNPPFVIHDMETLCMAEKTLVAKLVANGIQNKEAEVRIFHCCQCTSVETVTELTEFAKAIPGFANLDLNDQVTLLKYGVYEAIFAMLSSVMNKDGMLVAYGNGFITREFLKSLRKPFCDIMEPKFDFAMKFNALELDDSDISLFVAAIICCGDRPGLLNVGHIEKMQEGIVHVLRLHLQSNHPDDIFLFPKLLQKMADLRQLVTEHAQLVQIIKKTESDAALHPLLQEIYRDMY
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.5
  • Shipping temperature:
    4 °C
  • Immunogen:
    This PPARA antibody is generated from a mouse immunized with a recombination protein from the human region of human PPARA.
  • Purification:
    Protein G Purification
  • Pk:
    400 µl

Specifikace

O tomto produktu

Type: Primary
Antigen: PPARA
Clonality: Monoclonal
Clone: 1331CT894.186.143
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse