Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-IL4R Rabbit Polyclonal Antibody
  ABGEAP20570A
 :  Abgent
Anti-IL4R Rabbit Polyclonal Antibody
  ABGEAP20570A
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    Interleukin 4 Receptor
  • Antigen symbol:
    IL4R
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Gene ID:
    3566
  • Antigen synonyms:
    IL-4RA|IL4RA|CD124
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    89.658 kDa
  • Sequence:
    HLGLEPGEKVEDMPK
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.52
  • Shipping temperature:
    4 °C
  • Immunogen:
    This IL4R antibody is generated from a rabbit immunized with a KLH conjugated synthetic peptide between 677-691 amino acids from the C-terminal region of human IL4R.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: IL4R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human