Order Entry
Czech Republic
ContactUsLinkComponent
Anti-USF1 Mouse Monoclonal Antibody [clone: 1264CT170.274.14]
Anti-USF1 Mouse Monoclonal Antibody [clone: 1264CT170.274.14]
Catalog # ABGEAM2248B
Supplier:  Abgent
Anti-USF1 Mouse Monoclonal Antibody [clone: 1264CT170.274.14]
Catalog # ABGEAM2248B
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Upstream transcription factor 1
  • Antigen symbol:
    USF1
  • Clonality:
    Monoclonal
  • Clone:
    1264CT170.274.14
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    internal
  • Gene ID:
    7391
  • Antigen synonyms:
    FCHL1|UEF|MLTF|FCHL|bHLHb11|MLTFI|HYPLIP1
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    33.538 kDa
  • Sequence:
    0
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.41
  • Shipping temperature:
    4 °C
  • Immunogen:
    This USF1 antibody is generated from a mice immunized with a recombinant protein between 1-310 amino acids from the Central region of human USF1.
  • Purification:
    Protein G Purification
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: USF1
Clonality: Monoclonal
Clone: 1264CT170.274.14
Conjugation: Unconjugated
Epitope: internal
Host: Mouse
Isotype: IgG1
Reactivity: Human