Anti-CSH1 Rabbit Polyclonal Antibody
Catalog # ABGEAP20176A
Dodavatel: Abgent
Specifikace
- Antibody type:Primary
- Antigen symbol:CSH1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Amino acid number:42 to 70
- Storage buffer:pH 7,4
- Sequence:FEETYIPKDQKYSFC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.39
- Shipping temperature:4 °C
- Immunogen:This CSH1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 42-70 amino acids from the N-terminal region of human CSH1.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifikace
O tomto produktu
Type: Primary
Antigen: CSH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human