Anti-TIPRL Rabbit Polyclonal Antibody
ABGEAP18140B
: Abgent
- Antibody type:Primary
- Antigen name:TOR signaling pathway regulator
- Antigen symbol:TIPRL
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Gene ID:261726
- Antigen synonyms:TIP|TIP41
- Amino acid number:189 to 218
- Storage buffer:pH 7,4
- Molecular weight:31.444 kDa
- Sequence:EADKTYMLREYTSREC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:1.300
- Shipping temperature:4 °C
- Immunogen:This TIPRL antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 189-218 amino acids from the C-terminal region of human TIPRL.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Type: Primary
Antigen: TIPRL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human