Order Entry
Czech Republic
ContactUsLinkComponent
Anti-IgM µ Goat Polyclonal Antibody (CF™ 640R)
Anti-IgM µ Goat Polyclonal Antibody (CF™ 640R)
  20349-250UL
 :  Biotium
Anti-IgM µ Goat Polyclonal Antibody (CF™ 640R)
  20349-250UL
 :  Biotium

 

  • Antibody type:
    Secondary
  • Antigen symbol:
    IgM µ
  • Clonality:
    Polyclonal
  • Conjugation:
    CF® 640R
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Goat
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    50 μl
  • Pk:
    250 µl

 

 

CF™640R antibodies are affinity-purified antibodies labeled with far-red fluorescent CF™640R, a member of the novel CF™ dye series developed by Biotium for labeling proteins and other biomolecules. CF™ dyes are designed to offer advantages in brightness, photostability, and/or specificity compared to other commercial dyes, and are available in colors spanning the visible, far-red, and near-infrared spectra.

CF™640R is a far-red fluorescent dye with excitation/emission maxima at 642/662nm. CF™640R is spectrally similar to AlexaFluor®647, Cy®5, and DyLight®649. CF™640R is much brighter than Cy®5 and at least as bright as Alexa Fluor® 647 when excited at 633 or 635nm. However, the most important advantage of CF™640R is its exceptional photostability. CF™640R is far more photostable than the cyanine-based dyes Cy®5 and Alexa Fluor® 647, making it an excellent choice for demanding applications like confocal microscopy.

CF™ dyes are covered by pending U.S. and international patents. CF™ is a trademark of Biotium, Inc. Alexa Fluor® is a registered trademark of Molecular Probes, Inc. Cy® is a registered trademark of GE Healthcare. DyLight® is a registered trademark of Pierce Biotechnology.

 : Antibodies are supplied at 2mg/mL in PBS pH 7.4 containing 50% glycerol, 2mg/mL BSA (IgG- and protease-free) and 0.05% sodium azide. Highly cross-adsorbed antibodies are adsorbed against whole serum protein and have minimal cross-reactivity for multiple labeling experiments as indicated. Bv: bovine; Ch: chicken; Gt: goat; GP: Guinea pig; Hs: horse; Hu: human; Ms: mouse; Rb: rabbit; Sh: sheep; SHm: Syrian hamster; Sw: swine; Rt: rat.

Type: Secondary
Antigen: IgM MU CHAIN
Clonality: Polyclonal
Clone:
Conjugation: CF640R
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human