Order Entry
Czech Republic
ContactUsLinkComponent
Anti-DARPP-32 Rabbit Polyclonal Antibody
Anti-DARPP-32 Rabbit Polyclonal Antibody
Catalog # CAYM10603-100
Supplier:  Cayman Chemical
Anti-DARPP-32 Rabbit Polyclonal Antibody
Catalog # CAYM10603-100
Supplier:  Cayman Chemical

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    DARPP-32
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Rat
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Modification:
    phospho-Thr34
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    phosphopeptide corresponding to amino acid residues surrounding phospho-Thr34 of rat DARPP-32
  • Purification:
    purified by affinity chromatography
  • Size:
    1 Ea
  • Pk:
    100 µl

Specifications

About this item

Type: Primary
Antigen: DARPP-32
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat