Anti-IL25 Mouse Monoclonal Antibody (Biotin) [clone: 11K8B1]
Catalog # ROCK209-306-C58
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:interleukin 25
- Antigen symbol:IL25
- Clonality:Monoclonal
- Clone:11K8B1
- Conjugation:Biotin
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:No
- Format:Antibody fraction
- Form:Lyophilized
- Gene ID:64806
- Antigen synonyms:IL17E
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:Anti-IL-17E (MOUSE) Monoclonal Antibody was produced in mouse by repeated immunizations with mature full length recombinant human IL-17E produced in E.coli followed by hybridoma development.
- Purification:Protein G purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
This purified antibody has been tested for use in Flow Cytometry and western blotting. Specific conditions for reactivity should be optimized by the end user.
Recommended Dilutions: ELISA: 1:10000-1:50000; Flow Cytometry: 1µg/ml; Western Blot: 0.5µg/ml;
Restore with deionized water (or equivalent); 100 µl
Type: Primary
Antigen: IL25
Clonality: Monoclonal
Clone: 11K8B1
Conjugation: Biotin
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human