Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-SHANK1 Mouse Monoclonal Antibody (Unconjugated) [clone: S22-21]
  ROCK200-301-G43
undefined
Anti-SHANK1 Mouse Monoclonal Antibody (Unconjugated) [clone: S22-21]
  ROCK200-301-G43

 

  • Antibody type:
    Primary
  • Antigen name:
    SH3 and multiple ankyrin repeat domains 1
  • Antigen symbol:
    SHANK1
  • Clonality:
    Monoclonal
  • Clone:
    S22-21
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    Internal
  • Cross adsorption:
    No
  • Format:
    Antibody fraction
  • Form:
    Liquid (sterile filtered)
  • Gene ID:
    78957
  • Antigen synonyms:
    Sstrip|Spank1|Shank1a
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,250% (v/v) Glycerol
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Dry Ice
  • Immunogen:
    SHANK1 Antibody was produced in mice by repeated immunizations raised against a fusion Protein corresponding to an internal region of (SH3/PDZ domains) of rat Shank1.
  • Purification:
    Protein G purified
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Anti-SHANK1 Antibody is suitable for use in WB and IHC. Expect a band approximately ~190-220kDa (alternative splice variants) on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: Immunofluorescence: 1.0-10µg/ml; Western Blot: 1-10µg/ml; Immunochemistry: 0.1-1.0µg/ml

Type: Primary
Antigen: SHANK1
Clonality: Monoclonal
Clone: S22-21
Conjugation: Unconjugated
Epitope: Center
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Human, Rat