Anti-SHANK2 Mouse Monoclonal Antibody (Unconjugated) [clone: S23b-49]
Catalog # ROCK200-301-G42
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:SH3 and multiple ankyrin repeat domains 2
- Antigen symbol:SHANK2
- Clonality:Monoclonal
- Clone:S23b-49
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:Internal
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:171093
- Antigen synonyms:ProSAP1|Spank-3|CortBP1
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,250% (v/v) Glycerol
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:SHANK Antibody was produced in mice by repeated immunizations raised against a fusion Protein corresponding to an internal region of rat Shank2.
- Purification:Protein G purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Anti-SHANK Antibody is suitable for use in WB, IHC and IP. Expect a band approximately ~160kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.
Type: Primary
Antigen: SHANK2
Clonality: Monoclonal
Clone: S23b-49
Conjugation: Unconjugated
Epitope: Center
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Human, Rat