Anti-GRIN2B Mouse Monoclonal Antibody (Unconjugated) [clone: S59-36]
Catalog # ROCK200-301-G23
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:Glutamate receptor, ionotropic, N-methyl D-aspartate 2B
- Antigen symbol:GRIN2B
- Clonality:Monoclonal
- Clone:S59-36
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:24410
- Antigen synonyms:GluN2B
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,250% (v/v) Glycerol
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:NR2B Antibody was produced in mice by repeated immunizations raised against a fusion Protein corresponding near the extracellular N-terminus region of rat NR2B.
- Purification:Protein G purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Anti-NR2B Antibody is suitable for use in WB, IHC and IP. Expect a band approximately ~166kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.
Type: Primary
Antigen: GRIN2B
Clonality: Monoclonal
Clone: S59-36
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat