Order Entry
Czech Republic
ContactUsLinkComponent
Anti-GRIN2B Rabbit Polyclonal Antibody
Anti-GRIN2B Rabbit Polyclonal Antibody
Catalog # ROCK612-401-D92
undefined
Anti-GRIN2B Rabbit Polyclonal Antibody
Catalog # ROCK612-401-D92

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Glutamate receptor, ionotropic, N-methyl D-aspartate 2B
  • Antigen symbol:
    GRIN2B
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Rat
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Antibody fraction
  • Form:
    Liquid
  • Gene ID:
    24410
  • Antigen synonyms:
    GluN2B
  • Modification:
    phospho-Thr1252
  • Storage buffer:
    0,01 M HEPES, 0,15 M Sodium Chloride, pH 7,50,1 mg/ml Bovine Serum Albumin - IgG and Protease free, 50% (v/v) Glycerol
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Dry Ice
  • Immunogen:
    Anti-NMDA R2B pT1252 Antibody was producedby repeated immunizations with synthetic phospho-peptide corresponding to amino acid residues surrounding Tyr1252.
  • Purification:
    affinity-purified
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Anti-NMDA R2B pT1252 Antibody is suitable for use in Western Blotting and IHC. Expect a band of approximately 180 kDa in size corresponding to NMDAR NR2B subunit Protein phosphorylated at Tyr1252 in the appropriate cell lysate or extract.

Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:400;

Type: Primary
Antigen: GRIN2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat