Anti-Chorionic Gonadotropin, Human, beta Mouse Monoclonal Antibody [clone: HCGb/54]
Supplier: US Biological
Certificates
About this item
Recommended dilution: Immunochemistry: 0.5-1 ?g/ml
Type: Primary
Antigen: Chorionic Gonadotropin, Human, beta
Clonality: Monoclonal
Clone: HCGb/54
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Specifications
- Catalog No:
- USBI138531-20
- USBI138531-100
- Antigen symbol:
- CGB
- CGB
- Antigen name:
- Glycoprotein hormones, beta polypeptide
- Glycoprotein hormones, beta polypeptide
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- HCGb/54
- HCGb/54
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant protein corresponding to human hCG-beta.
- Recombinant protein corresponding to human hCG-beta.
- Form:
- liquid
- liquid
- Purification:
- purified by Protein A chromatography
- purified by Protein A chromatography
- Concentration:
- ~0,2 mg/ml
- ~0,2 mg/ml
- Cross adsorption:
- No
- No
- Storage buffer:
- PBS, 0,05% BSA, 0,05% sodium azide.
- PBS, 0,05% BSA, 0,05% sodium azide.
- Storage temperature:
- –20 °C
- –20 °C
Specifications
Frequently Bought Together