Order Entry
Czech Republic
ContactUsLinkComponent
Anti-IP-10 Rabbit Polyclonal Antibody
Anti-IP-10 Rabbit Polyclonal Antibody
Catalog # USBI143636-50
Supplier:  US Biological
Anti-IP-10 Rabbit Polyclonal Antibody
Catalog # USBI143636-50
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    C-X-C motif chemokine 10
  • Antigen symbol:
    CXCL10
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Rat
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Neutralisation:
    Yes
  • Form:
    lyophilized powder
  • Storage temperature:
    –20 °C
  • Immunogen:
    Highly purified rat IP-10.
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Recommended dilution: ELISA: 0.5-2?g/ml

Type: Primary
Antigen: IP-10
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat,