Anti-p53 Mouse Monoclonal Antibody [clone: DO-7]
Supplier: US Biological
Certificates
About this item
Recommended dilution: Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: p53
Clonality: Monoclonal
Clone: DO-7
Conjugation: unconjugated
Epitope: 20 to 25
Host: Mouse
Isotype: IgG2b
Reactivity: Human,
Specifications
- Catalog No:
- USBIP1001-26N5-25
- USBIP1001-26N5-100
- Antigen symbol:
- P53
- P53
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- DO-7
- DO-7
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG2b
- IgG2b
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Epitope:
- 20 to 25
- 20 to 25
- Immunogen:
- P53 N-terminal linear epitope aa20-25.
- P53 N-terminal linear epitope aa20-25.
- Form:
- liquid
- liquid
- Purification:
- purified by immunoaffinity chromatography
- purified by immunoaffinity chromatography
- Concentration:
- ~0,5 mg/ml
- ~0,5 mg/ml
- Cross adsorption:
- No
- No
- Storage buffer:
- PBS, pH 7,2, 0,09% sodium azide.
- PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:
- –20 °C
- –20 °C
Specifications
Frequently Bought Together