Anti-MHC Class 1, H2, KK Mouse Monoclonal Antibody [clone: 36-7-5]
Catalog # USBIM3885-85K-500
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:MHC Class 1, H2, KK
- Clonality:Monoclonal
- Clone:36-7-5
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Cross adsorption:No
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:–20 °C
- Concentration:~0,5 mg/ml
- Immunogen:A.AL mouse splenocytes
- Purification:purified by immunoaffinity chromatography
- Size:500 μg
- Pk:500 µG
Specifications
About this item
Type: Primary
Antigen: MHC Class 1, H2, KK
Clonality: Monoclonal
Clone: 36-7-5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Mouse,