Anti-CD45 Rat Monoclonal Antibody [clone: 30-F11]
Type: Primary
Antigen: CD45
Clonality: Monoclonal
Clone: 30-F11
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse,
- Catalog No:
- USBIC2400-20Q-50
- USBIC2400-20Q-500
- Antigen symbol:
- CD45
- CD45
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 30-F11
- 30-F11
- Reactivity:
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Isotype:
- IgG2b kappa
- IgG2b kappa
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Immunogen:
- Mouse thymus or spleen
- Mouse thymus or spleen
- Form:
- liquid
- liquid
- Purification:
- purified by immunoaffinity chromatography
- purified by immunoaffinity chromatography
- Concentration:
- ~0,5 mg/ml
- ~0,5 mg/ml
- Cross adsorption:
- No
- No
- Storage buffer:
- PBS, pH 7,2, 0,09% sodium azide.
- PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:
- –20 °C
- –20 °C
Frequently Bought Together