Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Helicobacter pylori Mouse Monoclonal Antibody [clone: 3/15/45/2]
Anti-Helicobacter pylori Mouse Monoclonal Antibody [clone: 3/15/45/2]
Supplier:  US Biological
Certificates
Anti-Helicobacter pylori Mouse Monoclonal Antibody [clone: 3/15/45/2]
Supplier:  US Biological

Specifications

  • Catalog No:
  • USBIH1840-03B-10
  • USBIH1840-03B-100
  • Antigen name:
  • Helicobacter pylori
  • Helicobacter pylori
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • 3/15/45/2
  • 3/15/45/2
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1
  • IgG1
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Helicobacter Pylori (H. Pylori)
  • Helicobacter Pylori (H. Pylori)
  • Form:
  • lyophilized powder
  • lyophilized powder
  • Purification:
  • purified by Protein G chromatography
  • purified by Protein G chromatography
  • Concentration:
  • ~1 mg/ml
  • ~1 mg/ml
  • Cross adsorption:
  • No
  • No
  • Storage buffer:
  • 50mM Tris, pH 7,2.
  • 50mM Tris, pH 7,2.
  • Storage temperature:
  • –20 °C
  • –20 °C

Specifications

Related Information

Frequently Bought Together