Anti-erbB Mouse Monoclonal Antibody [clone: EGF-R1]
Supplier: US Biological
Certificates
About this item
Type: Primary
Antigen: erbB
Clonality: Monoclonal
Clone: EGF-R1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human,
Specifications
- Catalog No:
- USBIE3375-35G-100
- USBIE3375-35G-10
- Antigen symbol:
- erbB
- erbB
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- EGF-R1
- EGF-R1
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Immunogen:
- Human EGF-Receptor
- Human EGF-Receptor
- Form:
- lyophilized powder
- lyophilized powder
- Purification:
- purified by Protein G chromatography
- purified by Protein G chromatography
- Concentration:
- ~1 mg/ml
- ~1 mg/ml
- Cross adsorption:
- No
- No
- Storage buffer:
- PBS, pH 7,2, 0,1% sodium azide.
- PBS, pH 7,2, 0,1% sodium azide.
- Storage temperature:
- –20 °C
- –20 °C
Specifications
Frequently Bought Together