Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-cyclic AMP Rabbit Polyclonal Antibody
Catalog # USBIC8450-12B-100
Supplier:  US Biological
Anti-cyclic AMP Rabbit Polyclonal Antibody
Catalog # USBIC8450-12B-100
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cyclic Adenosine Monophosphate
  • Antigen symbol:
    cAMP
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Format:
    Serum
  • Form:
    lyophilized powder
  • Storage temperature:
    –20 °C
  • Immunogen:
    Synthetic succinylated cAMP, BSA conjugated
  • Tested applications:
    Radio Immunoassay
  • Size:
    100 μl
  • Pk:
    100 µl

Specifications

About this item

Recommended dilution: Radio Immunoassay: 1:1000

Type: Primary
Antigen: cyclic AMP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: