Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-Brain Natriuretic Peptide Mouse Monoclonal Antibody [clone: 1-11-7]
Catalog # USBIB2702-34C-10
Supplier:  US Biological
Anti-Brain Natriuretic Peptide Mouse Monoclonal Antibody [clone: 1-11-7]
Catalog # USBIB2702-34C-10
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Brain Natriuretic Peptide
  • Antigen symbol:
    BNP
  • Clonality:
    Monoclonal
  • Clone:
    1-11-7
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Form:
    lyophilized powder
  • Amino acid number:
    1 to 21
  • Storage buffer:
    200mM Na Citrate-Tris, 500mM sodium chloride, pH 7,5, 0,02% sodium azide.
  • Storage temperature:
    –20 °C
  • Concentration:
    ~1 mg/ml
  • Immunogen:
    Synthetic human pro-BNP (aa 1-21) polylysinee conjugated (HPLGSPGSASDLETSGLQEQR)
  • Purification:
    purified by Protein A chromatography
  • Size:
    10 μg
  • Pk:
    10 µG

Specifications

About this item

Type: Primary
Antigen: Brain Natriuretic Peptide
Clonality: Monoclonal
Clone: 1-11-7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human,