Anti-Brain Natriuretic Peptide Mouse Monoclonal Antibody [clone: 3-56]
Catalog # USBIB2702-33C-10
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Brain Natriuretic Peptide
- Antigen symbol:BNP
- Clonality:Monoclonal
- Clone:3-56
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Cross adsorption:No
- Form:lyophilized powder
- Amino acid number:1 to 32
- Storage buffer:PBS, pH 7,2
- Storage temperature:–20 °C
- Concentration:~1 mg/ml
- Immunogen:Synthetic human BNP (aa 1-32) KLH conjugated Synonyms: Brain natriuretic peptide 32, BNP-32
- Purification:purified by Protein G chromatography
- Size:10 μg
- Pk:10 µG
Specifications
About this item
Type: Primary
Antigen: Brain Natriuretic Peptide
Clonality: Monoclonal
Clone: 3-56
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human,