Order Entry
Czech Republic
ContactUsLinkComponent
Anti-BD1 Mouse Monoclonal Antibody [clone: M4-14b-H4]
Anti-BD1 Mouse Monoclonal Antibody [clone: M4-14b-H4]
Catalog # USBIB0899-03G-10
Supplier:  US Biological
Anti-BD1 Mouse Monoclonal Antibody [clone: M4-14b-H4]
Catalog # USBIB0899-03G-10
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    BD1
  • Clonality:
    Monoclonal
  • Clone:
    M4-14b-H4
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    lyophilized powder
  • Amino acid number:
    1 to 36
  • Storage buffer:
    50mM Tris, pH 7,4.
  • Storage temperature:
    –20 °C
  • Concentration:
    ~1 mg/ml
  • Immunogen:
    Synthetic human β-Defensin 1 (aa 1-36) (3 disulfide bridges) (DHYNCVSSGGQCLYSACPIFTKIQGTCYRGKAKCCK)
  • Purification:
    purified by Protein G chromatography
  • Size:
    10 μg
  • Pk:
    10 µG

Specifications

About this item

Recommended dilution: Western Blot: 1:1000-1:2000

Type: Primary
Antigen: BD1
Clonality: Monoclonal
Clone: M4-14b-H4

Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human,