Order Entry
Czech Republic
ContactUsLinkComponent
Anti-IP-10 Mouse Monoclonal Antibody [clone: 8A201]
Anti-IP-10 Mouse Monoclonal Antibody [clone: 8A201]
Catalog # USBII8449-05B-100
Supplier:  US Biological
Anti-IP-10 Mouse Monoclonal Antibody [clone: 8A201]
Catalog # USBII8449-05B-100
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    C-X-C motif chemokine 10
  • Antigen symbol:
    CXCL10
  • Clonality:
    Monoclonal
  • Clone:
    8A201
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,4, 0,02% sodium aide.
  • Storage temperature:
    –20 °C
  • Concentration:
    ~1 mg/ml
  • Immunogen:
    Purified recombinant human IP-10 expressed in PET32a produced in E. coli.
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 μg
  • Pk:
    100 µG

Specifications

About this item

Recommended dilution: ELISA: 0.1-1.0 ?g/ml; Western Blot: 1-2?g/ml

Type: Primary
Antigen: IP-10
Clonality: Monoclonal
Clone: 8A201
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human,