Anti-Interleukin 17F Rabbit Polyclonal Antibody
Catalog # USBII8439-28-100
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 17F
- Antigen symbol:IL17F
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:lyophilized powder
- Storage buffer:50% glycerol, dH2O to 0,1-1mg/ml.
- Storage temperature:–20 °C
- Concentration:~1 mg/ml
- Immunogen:Highly pure (> 98%) recombinant human IL-17fF
- Purification:purified by immunoaffinity chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA: 0.5-2?g/ml; Western Blot: 01-02?g/ml
Type: Primary
Antigen: Interleukin 17F
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,