Anti-Fibroblast Growth Factor 16 Goat Polyclonal Antibody (Biotin)
Supplier: US Biological
Certificates
About this item
Recommended dilution: ELISA: 0.15-0. 30 ?g/ml
Type: Primary
Antigen: Fibroblast Growth Factor 16
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human,
Specifications
- Catalog No:
- USBIF4212-50B1-25
- USBIF4212-50B1-50
- Antigen symbol:
- FGF16
- FGF16
- Antigen name:
- Fibroblast growth factor 16
- Fibroblast growth factor 16
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Goat
- Goat
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Highly pure (> 98%) recombinant human FGF-16
- Highly pure (> 98%) recombinant human FGF-16
- Form:
- lyophilized powder
- lyophilized powder
- Purification:
- purified by immunoaffinity chromatography
- purified by immunoaffinity chromatography
- Concentration:
- ~0,05 mg/ml
- ~0,05 mg/ml
- Cross adsorption:
- No
- No
- Storage buffer:
- PBS
- PBS
- Storage temperature:
- –20 °C
- –20 °C
Specifications
Frequently Bought Together