Anti-Interferon Stimulating Gene 15 Mouse Monoclonal Antibody [clone: 7G32]
Catalog # USBII7662-94B-100
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Interferon Stimulating Gene 15
- Clonality:Monoclonal
- Clone:7G32
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,1% sodium azide.
- Storage temperature:–20 °C
- Concentration:~1 mg/ml
- Immunogen:Recombinant human ISG15 (1-157aa) (E. coli).
- Purification:purified by Protein G chromatography
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:1000-1:2000
Type: Primary
Antigen: Interferon Stimulating Gene 15
Clonality: Monoclonal
Clone: 7G32
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human,