Anti-ZNF15 Rabbit Polyclonal Antibody
Supplier: US Biological
Certificates
About this item
Recommended dilution: ELISA:1:175000
Type: Primary
Antigen: ZNF15
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,
Specifications
- Catalog No:
- USBIZ0737-15-50
- USBIZ0737-15-100
- Antigen symbol:
- ZNF15
- ZNF15
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide KIRRHKKEKEEKKRKKP, corresponding to human ZNF15 ((KLH). MW: 218.4kD.
- Synthetic peptide KIRRHKKEKEEKKRKKP, corresponding to human ZNF15 ((KLH). MW: 218.4kD.
- Sequence:
- KIRRHKKEKEEKKRKKP
- KIRRHKKEKEEKKRKKP
- Form:
- liquid
- liquid
- Purification:
- purified by immunoaffinity chromatography
- purified by immunoaffinity chromatography
- Concentration:
- ~0,6mg/ml
- ~0,6mg/ml
- Cross adsorption:
- No
- No
- Storage buffer:
- PBS, 0,02% sodium azide, before the addition of glycerol to 40%.
- PBS, 0,02% sodium azide, before the addition of glycerol to 40%.
- Storage temperature:
- –20 °C
- –20 °C
Specifications
Frequently Bought Together