Anti-Splicing Factor, Arginine/Serine-Rich 14 Rabbit Polyclonal Antibody
Catalog # USBIS5554-78-100
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Splicing Factor, Arginine/Serine-Rich 14
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rooster,Human
- Cross adsorption:No
- Form:liquid
- Storage buffer:PBS, 0,02% sodium azide, before the addition of glycerol to 40%.
- Sequence:SLPDRNDAAKDCPP
- Storage temperature:–20 °C
- Concentration:~0,2 mg/ml
- Immunogen:Synthetic peptide, SLPDRNDAAKDCPP corresponding to human SFRS14 (KLH).
- Purification:purified by immunoaffinity chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA: 1:75000
Type: Primary
Antigen: Splicing Factor, Arginine/Serine-Rich 14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rooster,dent,