Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Splicing Factor, Arginine/Serine-Rich 10 Rabbit Polyclonal Antibody
Anti-Splicing Factor, Arginine/Serine-Rich 10 Rabbit Polyclonal Antibody
Catalog # USBIS5554-70-100
Supplier:  US Biological
Anti-Splicing Factor, Arginine/Serine-Rich 10 Rabbit Polyclonal Antibody
Catalog # USBIS5554-70-100
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Splicing Factor, Arginine/Serine-Rich 10
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Rooster,
    Human
  • Cross adsorption:
    No
  • Form:
    liquid
  • Storage buffer:
    PBS, 0,02% sodium azide, before the addition of glycerol to 40%.
  • Molecular weight:
    33.67 kDa
  • Storage temperature:
    –20 °C
  • Concentration:
    ~1 mg/ml
  • Immunogen:
    Synthetic peptide: RSYSRDYRRRHSHS (KLH).
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    100 μg
  • Pk:
    100 µG

Specifications

About this item

Recommended dilution: ELISA: 1:112000

Type: Primary
Antigen: Splicing Factor, Arginine/Serine-Rich 10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rooster,dent,