Anti-AP2 Rabbit Polyclonal Antibody
Catalog # AGRIAS122609
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen name:Floral homeotic protein APETALA 2
- Antigen symbol:AP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Echinochla crus-galli,Spinacia oleracea,Schima superba,A. thaliana,Manihot esculenta,Digitaria sanguinalis,Silene vulgaris,Liquidambar formosana,N. tabacum thylakoid-bound apX
- Western blot:Yes
- Cross adsorption:No
- UniProtKB:P47927
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated synthetic peptide derived from AP2 sequence of Arabidopsis thaliana,
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: AP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, Digitaria sanguinalis, Echinochla crus-galli, Liquidambar formosana, Manihot esculenta, N. tabacum thylakoid-bound apX, Schima superba, Silene vulgaris, Spinacia oleracea