Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-3-nitroY Mouse Monoclonal Antibody
  AGRIAS10706-100
 :  Agrisera
Anti-3-nitroY Mouse Monoclonal Antibody
  AGRIAS10706-100
 :  Agrisera

 

  • Antibody type:
    Primary
  • Antigen name:
    3-(4-Hydroxy-3-nitrophenylacetamido) propionic acid
  • Antigen symbol:
    3-nitroY
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Human,
    Dog,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ambient
  • Immunogen:
    BSA-conjugated 3-(4-Hydroxy-3-nitrophenyl acetamido) propionic acid
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

Type: Primary
Antigen: 3-nitroY
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Mouse, Rat