Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-IgG (H+L) Rabbit Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
  AGRIAS101101
 :  Agrisera
Anti-IgG (H+L) Rabbit Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
  AGRIAS101101
 :  Agrisera

 

  • Antibody type:
    Secondary
  • Antigen symbol:
    IgG (H+L)
  • Clonality:
    Polyclonal
  • Conjugation:
    TRITC (Tetramethylrhodamine Isothiocyanate)
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Rabbit,
    Human,
    Bovine,
    Mouse
  • Cross adsorption:
    No
  • Storage temperature:
    +2°C to +8 °C
  • Shipping temperature:
    Ambient
  • Immunogen:
    Purified Goat IgG, whole molecule
  • Purification:
    purified
  • Size:
    1 mg
  • Pk:
    1 mg

 

 

Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Human, Mouse, Rabbit