Anti-c-Myc Chicken Polyclonal Antibody
Catalog # AGRIAS09601
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:C-MYK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Chicken
- ImmunoChemistry:Yes
- Reactivity:Synechococcus sp.,C. reinhardtii,A. thaliana,Streptomyces sp.
- Epitope:C-terminal
- Cross adsorption:No
- Amino acid number:408 to 420
- Sequence:MEQKLISEEDLNE
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated peptide, amino acids 408-420 ( sequence MEQKLISEEDLNE), of the C-terminal domian of human c-Myc P01106.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: C-MYK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Chicken
Isotype:
Reactivity: A. thaliana, C. reinhardtii, Streptomyces sp., Synechococcus sp.