Anti-N6-isopentenyladenosine Rabbit Polyclonal Antibody
Catalog # AGRIAS09435
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen name:N6-isopentenyladenosine
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:BSA-conjugated, via ribose, N6-isopentenyladenosine
- Tested applications:Immunolocalization
- Size:200 µg
- Pk:200 µG
Specifications
About this item
Type: Primary
Antigen: N6-isopentenyladenosine
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: