Anti-meta-topolin riboside Rabbit Polyclonal Antibody
Catalog # AGRIAS09417
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen name:meta-topolin riboside
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:BSA-conjugated, via ribose, meta-topolin riboside
- Tested applications:Radio Immunoassay
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Primary
Antigen: meta-topolin riboside
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: