Anti-L-30 Rabbit Polyclonal Antibody
Catalog # AGRIAS08331
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:L-30
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:A. thaliana
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:purified native Chlamydomonas reinhardtii L-30 protein eluted from a gel piece
- Purification:purified
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: L-30
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana