Anti-H+ATPase Rabbit Polyclonal Antibody
AGRIAS07260
: Agrisera
- Antibody type:Primary
- Antigen name:H+ATPase
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Cucurbita moschata,Populus tremula,H. vulgare,Soy bean,Noccaea caerulescens,Medicago truncatula,Pteris vittata,Oryza sativa,Spinacia oleracea,Picea abies,C. reinhardtii,Ricinus communis,Phalenopsis sogo yukidian cultivar v3,Kandelia obovata,Zea mays,A. thaliana,N. tabacum,Solanum lycopersicum
- Western blot:Yes
- Cross adsorption:No
- UniProtKB:Q9LV11
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated synthetic peptide exposed to cytoplasm in H+ATPase model, derived from available di and monocot, fern, mosses and algal plasma membrane ATPase sequences including Arabidopsis thaliana ATPase 1 (
- Tested applications:Immunolocalization
- Size:50 µl
- Pk:50 µl
Type: Primary
Antigen: H+ATPase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, C. reinhardtii, Cucurbita moschata, Glycine max, H. vulgare, Kandelia obovata, Solanum lycopersicum, Medicago truncatula, N. tabacum, Noccaea caerulescens, Oryza sativa, Phalenopsis sogo Yukidian cultivar v3, Picea abies, Populus tremula, Pteris vittata, Ricinus communis, Spinacia oleracea, Zea mays