Anti-Fdx3 Rabbit Polyclonal Antibody
Catalog # AGRIAS07224
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:FDX3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Chlamydomonas reinhardtii
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated synthetic peptide chosen from a C-terminal of Chlamydomonas reinhardtii Q2HZ24
- Size:200 µg
- Pk:200 µG
Specifications
About this item
Type: Primary
Antigen: FDX3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Chlamydomonas reinhardtii