Anti-HSF1 Rabbit Polyclonal Antibody
Catalog # AGRIAS06178
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:HSF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Cucurbita pepo,Pisum sativum,Triticum sp.
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:No
- Antigen synonyms:HSP70
- UniProtKB:A1YTN3
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:C-terminal part of Chlamydomonas reinhardtii HSF1 (Exons 7 to 9),
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: HSF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Cucurbita pepo, Pisum sativum, triticum sp.