Anti-hScrib Rabbit Polyclonal Antibody
Catalog # AGRIAS06155
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:HSCRIB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Salmon
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:GST tagged C-terminus of hScrib Q14160 including PDZ3 and 4 domains
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: HSCRIB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Salmon