Anti-ANO1 Mouse Monoclonal Antibody [clone: 2F4]
Catalog # ORIGTA803607
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Anoctamin 1, calcium activated chloride channel
- Antigen symbol:ANO1
- Clonality:Monoclonal
- Clone:2F4
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:TMEM16A
- Amino acid number:1 to 333
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Human recombinant protein fragment corresponding to amino acids 1-333 of human ANO1 (NP_060513) produced in E.coli.
- Purification:purified by affinity chromatography
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:2000
Type: Primary
Antigen: ANO1
Clonality: Monoclonal
Clone: 2F4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human