Anti-ANO1 Mouse Monoclonal Antibody [clone: 1H2]
Catalog # ORIGTA803368
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Anoctamin 1, calcium activated chloride channel
- Antigen symbol:ANO1
- Clonality:Monoclonal
- Clone:1H2
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:TMEM16A
- Amino acid number:1 to 333
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Human recombinant protein fragment corresponding to amino acids 1-333 of human ANO1 (NP_060513) produced in E.coli.
- Purification:purified by affinity chromatography
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Immunochemistry: 1:150
Type: Primary
Antigen: ANO1
Clonality: Monoclonal
Clone: 1H2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human