Anti-TP53 Mouse Monoclonal Antibody [clone: DO-1]
Catalog # ORIGTA804804
Supplier: OriGene
Specifications
- Druh protilátky:Primární
- Symbol antigenu:TP53
- Klonovalita:Monoclonal
- Klon:DO-1
- Spojení:Unconjugated
- Hostitel:Mouse
- Imunochemie:Yes
- Izotyp:IgG2a
- Reaktivita:Human
- Western blot:Yes
- Křížová adsorbce:No
- Formulář:Liquid
- Skladovací pufr:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Skladovací teplota:–20 °C
- Transportní teplota:–20 °C
- Imunogen:Recombinant human wild type p53 protein expressed in E. coli.
- Čištění:purified by affinity chromatography
- Rozměry protilátky:100 μl
- Bal.:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150
Type: Primary
Antigen: TP53
Clonality: Monoclonal
Clone: DO-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human