Order Entry
Czech Republic
ContactUsLinkComponent
Anti-TRPC7 Mouse Monoclonal Antibody [clone: S64A-36]
Anti-TRPC7 Mouse Monoclonal Antibody [clone: S64A-36]
Catalog # ORIGTA326454
Supplier:  OriGene
Anti-TRPC7 Mouse Monoclonal Antibody [clone: S64A-36]
Catalog # ORIGTA326454
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    TRPC7
  • Clonality:
    Monoclonal
  • Clone:
    S64A-36
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Amino acid number:
    845 to 862
  • Storage buffer:
    PBS pH 7,4, 50% glycerol, 0,09% sodium azide
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Synthetic peptide amino acids 845-862 of human TRPC7,
  • Purification:
    purified by Protein G chromatography
  • Size:
    100µg
  • Pk:
    100 µG

Specifications

About this item

Recommended Dilutions: Western Blot: 1-10?g/ml; Immunochemistry: 0.1-1.0?g/ml; Immunofluorescence: 1.0-10?g/ml

Type: Primary
Antigen: TRPC7
Clonality: Monoclonal
Clone: S64A-36
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat