Order Entry
Czech Republic
ContactUsLinkComponent
Anti-SLC18A3 Mouse Monoclonal Antibody [clone: S6-38]
Anti-SLC18A3 Mouse Monoclonal Antibody [clone: S6-38]
Catalog # ORIGTA326422
Supplier:  OriGene
Anti-SLC18A3 Mouse Monoclonal Antibody [clone: S6-38]
Catalog # ORIGTA326422
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Solute carrier family 18, member 3
  • Clonality:
    Monoclonal
  • Clone:
    S6-38
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Amino acid number:
    521 to 532
  • Storage buffer:
    PBS pH 7,4, 50% glycerol, 0,09% sodium azide.
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Synthetic peptide amino acids 521-532 of human VACht
  • Purification:
    purified by Protein G chromatography
  • Size:
    100µg
  • Pk:
    100 µG

Specifications

About this item

Recommended Dilutions: Immunochemistry: 1:50-1:200

Type: Primary
Antigen: SLC18A3
Clonality: Monoclonal
Clone: S6-38
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat