Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-MSH6 Rabbit Polyclonal Antibody
  ORIGTA323720
 :  OriGene
Anti-MSH6 Rabbit Polyclonal Antibody
  ORIGTA323720
 :  OriGene

 

  • Antibody type:
    Primary
  • Antigen symbol:
    MSH6
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Storage buffer:
    PBS pH 7,3, 0,05% NaN3, 50% glycerol
  • Molecular weight:
    153 kDa
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Fusion protein corresponding to C terminal 300 amino acids of human mutS homolog 6 (E. coli)
  • Purification:
    purified by antigen affinity chromatography
  • Size:
    100 μl
  • Pk:
    100 µl

 

 

Recommended Dilutions: ELISA: 1:1000-2000; Western Blot: 1:200-1000; Immunochemistry: 1:10-50

Type: Primary
Antigen: MSH6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat